FibrOut™, Mouse 1 Fibroblast Growth Inhibitors, CHI Scientific
Supplier: CHI Scientific
4-21501 4-21502
10785-982EA 669.51 CAD
10785-982 10785-984
FibrOut™, Mouse 1 Fibroblast Growth Inhibitors, CHI Scientific
Cell Culture Supplements and Additives
The FibrOutTM System is a combination of several biochemical compounds and reagents which prevent fibroblastic overgrowth and increase targeted cell growth during primary cell culture
The reagents contained in this system are stable at room temperatures for normal shipping periods, but the package should be stored at 4oC upon arrival. The reagent is stable and effective for up to 4 months at 4oC. For long periods of storage, it should be kept at -20oC. Avoid repeated freeze-thaw cycles.
Each fibroblast growth-inhibition system is customized to be tissue-specific and cell-specific thereby producing the maximum amount of target cells. Each system contains sufficient materials for making 500 ml of cell culture medium. Each lot is tested for fibroblast growth inhibition performance and is composed of freshly prepared reagent solutions.
Order Now
SPECIFICATIONS
Environmentally Preferable
Species
Source
Learn more