Anti-capsaicinR Rabbit Polyclonal Antibody
Supplier: Biosensis
R-076-100
10782-700EA 542.49 CAD
10782-700
Anti-capsaicinR Rabbit Polyclonal Antibody
Antibodies
Responses evoked by low pH and heat, and capsaicin can be antagonized by capsazepine. SIMILARITY: Belongs to the transient receptor family. TrpV subfamily. SIMILARITY: Contains 3 ANK repeats.
Specificity was confirmed by IHC using frozen sections of rat dorsal root ganglia (DRG) and spinal cord. Due to sequence homology, similar staining is predicted in human DRG and spinal cord.
Application Information:
IHC. Use at 1:1000 to 1:2000 dilution. This antibody has not been tested in other applications. Biosensis recommends optimal dilutions/concentrations should be determined by the end user.
More Product Information