Anti-rh NTN Rabbit Polyclonal Antibody
Supplier: Biosensis
R-1561-100
10782-534EA 559.08 CAD
10782-534
Anti-rh NTN Rabbit Polyclonal Antibody
Antibodies
Neurturin (NTN) is a member of the GDNF family of neurotrophic factors. This protein is a potent survival factor for several populations of central and peripheral neurons in mature and developing rodents. FUNCTION: Supports the survival of sympathetic neurons in culture. May regulate the development and maintenance of the CNS. Might control the size of non-neuronal cell population such as haemopoietic cells. SUBUNIT: Homodimer; disulfide-linked. SUBCELLULAR LOCATION: Secreted protein. DISEASE: Defects in NRTN are a cause of Hirschsprung disease (HSCR). In association with mutations of RETMore Product Information