Anti-H3 Rabbit Polyclonal Antibody

Supplier: Biosensis
R-1549-100
10782-518EA 559.08 CAD
10782-518
Anti-H3 Rabbit Polyclonal Antibody
Antibodies
Histone H3 is a core component of nucleosome.

Antibody binds to various Histone H3 proteins.
Application Information:
WB. Suggested dilution of 1:1,000-1:5,000. Biosensis recommends that the optimal working dilution should be determined by the end user.

Type: Primary
Antigen: Histone H3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-terminus of Human Histone H3 (121-136 aa).
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat, Bovine
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR