Anti-Neurofilament Light Rabbit Polyclonal Antibody
Supplier: Biosensis
R-1392-50
10782-464EA 542.49 CAD
10782-464
Anti-Neurofilament Light Rabbit Polyclonal Antibody
Antibodies
Neurofilaments are composed of three intermediate filament proteins: light (~68 kDa), medium (~160 kDa) and heavy (~200 kDa), which are involved in the maintenance of the neuronal caliber. Neurofilament light (NF68 or NF-L) is the most abundant of the three proteins.
Specifically recognizes the light neurofilament subunit NF-L (~70 kDa) in WB.
Application Information:
Western Blotting (WB), Immunocytochemistry (ICC) and Immunohistochemistry (IHC). A dilution of 1:10,000 - 1:20,000 is recommended for WB. A dilution of 1:2,000 - 1:5,000 is recommended for ICC/IHC. Biosensis recommends optimalMore Product Information