Anti-Neurofilament Heavy, phosphorylated and non-phosphorylated Rabbit Polyclonal Antibody
Supplier: Biosensis
R-1389-50
10782-462EA 542.49 CAD
10782-462
Anti-Neurofilament Heavy, phosphorylated and non-phosphorylated Rabbit Polyclonal Antibody
Antibodies
Neurofilaments contain three intermediate filament proteins: light (68 kDa), medium (160 kDa) and heavy (200 kDa). Neurofilament heavy (NF200 or NF-H) is phosphorylated and it is thought that this results in the formation of interfilament cross bridges that are important in the maintenance of axonal caliber.
The specificity of this antibody has been confirmed by WB. This antibody reacts with noth phosphorylated NF-H at approx 200 kDa and non-phosphorylated NF-H at 160 kDa.
Application Information:
Western Blotting (WB) and Immunocytochemistry (IC). Suggested dilution for WB of 1:5,000-10,00More Product Information