Anti-Neurofilament Heavy, phosphorylated Rabbit Polyclonal Antibody
Supplier: Biosensis
R-1388-50
10782-460EA 542.49 CAD
10782-460
Anti-Neurofilament Heavy, phosphorylated Rabbit Polyclonal Antibody
Antibodies
Neurofilaments contain three intermediate filament proteins: light (68 kDa), medium (160 kDa) and heavy (200 kDa). Neurofilament heavy (NF200 or NF-H) is phosphorylated and it is thought that this results in the formation of interfilament cross bridges that are important in the maintenance of axonal caliber.
The specificity of this antibody has been confirmed by WB. This antibody reacts with phosphorylated NF-H and is seen as a band of approx 200 kDa.
Application Information:
Western Blotting (WB), Immunocytochemistry (ICC) and Immunocytochemistry (IHC). Suggested dilution for WB of 1:5,000-More Product Information