Anti-GFRA2 Rabbit Polyclonal Antibody
Supplier: Biosensis
R-067-100
10782-362EA 559.08 CAD
10782-362
Anti-GFRA2 Rabbit Polyclonal Antibody
Antibodies
FUNCTION: Receptor for neurturin. Mediates the NRTN-induced autophosphorylation and activation of the RET receptor. Also able to mediate GDNF signaling through the RET tyrosine kinase receptor. SUBCELLULAR LOCATION: Cell membrane; lipid-anchor; GPI-anchor (By similarity). ALTERNATIVE PRODUCTS: 2 named isoforms produced by alternative splicing.
Western blot analysis indicates a high level of specificity to GFR alpha-2 for this antiserum.
Application Information:
IHC, WB. A dilution of 1:500 to 1:1000 is recommended for both applications. This antibody stains beautifully the large neurons inMore Product Information