Anti-V5 Tag Mouse Monoclonal Antibody [clone: BE10]

Supplier: Biosensis
M-1317-100
10782-054EA 625.44 CAD
10782-054
Anti-V5 Tag Mouse Monoclonal Antibody [clone: BE10]
Antibodies
The V5 epitope corresponds to a region from Simian Virus Type 5 (SV5).

Antibody detects V5-tagged fusion protein.
Application Information:
Western Blotting (WB), Immunohistochemistry (IHC) and ELISA. Suggested dilutions for WB at 1:500 - 1:1000, ELISA at 1:5,000-10,000 and IHC at 1:50-1:100. Biosensis recommends optimal dilutions/concentrations should be determined by the end user.

Type: Tag
Antigen: V5 Tag
Clonality: Monoclonal
Clone: BE10
Conjugation: Unconjugated
Epitope: GKPIPNPLLGLDST
Host: Mouse
Isotype:
Reactivity:
Order Now

SPECIFICATIONS

Antigen symbol V5 Tag
Antigen name V5 Epitope Tag
Environmentally Preferable
Antibody type Tag
Clonality Monoclonal
Conjugation Unconjugated
Clone BE10
Reactivity
Host Mouse
Isotype
Western blot Yes
ELISA Yes
ImmunoChemistry Yes
Epitope GKPIPNPLLGLDST
Immunogen Synthetic peptide GKPIPNPLLGLDST
Format Liquid
Cross adsorption No

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR