Anti-Red Fluorescent Protein Mouse Monoclonal Antibody [clone: B25]
Supplier: Biosensis
M-1315-100
10782-052EA 625.44 CAD
10782-052
Anti-Red Fluorescent Protein Mouse Monoclonal Antibody [clone: B25]
Antibodies
Designed to detect Red Fluorescent Protein (RFP) and its variants in ELISA (sandwich or capture), immunoblotting, immunoprecipitation and immunohistochemistry.
Designed to detect Red Fluorescent Protein (RFP) and its variants in ELISA (sandwich or capture), immunoblotting, immunoprecipitation and immunohistochemistry. Western Blotting (WB), Immunohistochemistry (IHC) and Immunoprecipitation (IP). Suggested starting dilutions are as follows: WB at 1:1,000, IP at 5 µg/mg lysate and IHC at 1:200. Biosensis recommends optimal dilutions/concentrations should be determined by the end user.
More Product Information