Anti-HA Tag Mouse Monoclonal Antibody [clone: BHAC5]

Supplier: Biosensis
M-1309-100
10782-046EA 625.44 CAD
10782-046
Anti-HA Tag Mouse Monoclonal Antibody [clone: BHAC5]
Antibodies
The Human influenza hemagglutin (HA) tag corresponds to a region (98-106 amino acids) from the HA molecule.

Antibody detects HA-tagged proteins (on amino- and carboxy termini) in transfected mammalian cells.
Application Information:
Western Blotting (WB), Immunoprecipitation (IP) and Immunohistochemistry (IH). Suggested dilutions for WB of 1:1000, IP and IS at 1:200. Biosensis recommends optimal dilutions/concentrations should be determined by the end user.

Type: Tag
Antigen: HA Tag
Clonality: Monoclonal
Clone: BHAC5
Conjugation: Unconjugated
Epitope: HA-tag YPYDVPDYA
Host: Mouse
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR