Human Recombinant sCD27 (from CHO Cells)
Supplier: Peprotech
310-30-10UG 310-30-50UG 310-30-100UG 310-30-250UG 310-30-500UG 310-30-1MG
10780-654EA 209.84 CAD
10780-654 10780-656 10780-658 10780-660 10780-662 10780-664
Human Recombinant sCD27 (from CHO Cells)
Proteins and Peptides
CD27 Ligand, a type II transmembrane protein, is a member of the TNF superfamily. It is expressed on activated T and B lymphocytes, as well as NK cells. CD27L and its receptor (CD27) regulate the immune response by promoting T-cell expansion and differentiation, as well as NK enhancement. CD27 signaling can act as a co-stimulatory effector to sustain the survival of CD8+ T cells, primarily by inducing increased expression of the IL-2 gene. Full length human CD27L is a 193 amino acid protein, consisting of a 17 amino acid cytoplasmic domain, a 21 amino acid transmembrane domain, and a 155 amino acid extracellular domain. Human soluble CD27L corresponds to the 155 amino acid extracellular domain of the full length CD27L protein. PeproTech’s Recombinant Human sCD27 Ligand contains the extracellular domain plus an N-terminal His-Tag. The calculated molecular weight of Recombinant Human sCD27 Ligand is 18.8 kDa.
Ordering information: For research use only. Not for use in diagnostic or therapeutic procedures.