Human Recombinant IFN-?2 (from E. coli)

Supplier: Peprotech

300-02K-5UG 300-02K-20UG 300-02K-100UG 300-02K-250UG 300-02K-500UG 300-02K-1MG
10780-372EA 209.84 CAD
10780-372 10780-374 10780-376 10780-378 10780-380 10780-382
Human Recombinant IFN-?2 (from E. coli)
Proteins and Peptides

IFNλ1, 2, and 3 (also known as IL-29, IL-28A and IL-28B respectively) are distantly related to the IL-10 family and interferons. All three IFN-lambdas use a distinct receptor system composed of an IFN-λR1 subunit (also called CRF2-12) and IL-10R2 subunit (also called CRF2-14). Signaling through this receptor system induces antiviral defenses similar to, but distinct from, than that of type I interferons. Recombinant Human IFN-λ2 is a 19.6 kDa protein containing 174 amino acid residues.


Ordering information: For research use only. Not for use in diagnostic or therapeutic procedures.

Order Now

SPECIFICATIONS

Protein/peptide name IFN-lambda 2
Environmentally Preferable
Protein synonyms IL-28A
Protein/peptide type Recombinant
Protein Function Cytokines and Growth Factors
Species Human
Source E. coli
Conjugation Unconjugated
Gene ID 282616 Q8IZJ0
Sequence PVARLHGALP DARGCHIAQF KSLSPQELQA FKRAKDALEE SLLLKDCRCH SRLFPRTWDL RQLQVRERPM ALEAELALTL KVLEATADTD PALVDVLDQP LHTLHHILSQ FRACIQPQPT AGPRTRGRLH HWLYRLQEAP KKESPGCLEA SVTFNLFRLL TRDLNCVASG DLCV
Protease-free Y
Endotoxin level Low
Endotoxin content >0.1 ng/μg
Biological activity Biological Activity tests can vary. Please refer to the Product CoA for information.
Purity 0.98
Form Lyophilized
Formulation Formulation can vary. Please refer to product CoA for information.
Molecular weight 19.6 kDa
Reconstitution instructions Reconstitution instructions can vary. Please refer to the product CoA for info.
Storage conditions Store at –20 °C prior to reconstitution.
Shipping temperature Ambient

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR