Human GRO-α/MGSA (CXCL1) Standard ABTS ELISA Development Kit, PeproTech, Inc.
Supplier: Peprotech
900-K38
10777-676KT 911.83 CAD
10777-676
Human GRO-α/MGSA (CXCL1) Standard ABTS ELISA Development Kit, PeproTech, Inc.
Assays ELISAs
PeproTech's Human GRO-α/MGSA (CXCL1) ELISA development kit contains the key components required for the quantitative measurement of natural and/or recombinanth human GRO-α/MGSA (CXCL1) in a sandwich ELISA format.
  • Easy-to-use
  • Optimized reagents for singleplex protein detection by solid phase sandwich enzyme-linked immunosorbent assay (ELISA)
Human GRO-α/MGSA (CXCL1) ELISA development kit contains the key components required for the quantitative measurement of natural and/or recombinanth human GRO-α/MGSA (CXCL1) in a sandwich ELISA format. Using the ELISA protocol, the recommended microplates, reagents and solutions, the components supplied in this kit are sufficient to assay human GRO-α/MGSA (CXCL1) in approximately 1000 ELISA plate wells.