Anti-Eotaxin (CCL11) Goat Polyclonal Antibody (Biotin)

Supplier: Peprotech
500-P41GBT-25UG 500-P41GBT-50UG
10775-296EA 536.88 CAD
10775-296 10775-298 10775-300
Anti-Eotaxin (CCL11) Goat Polyclonal Antibody (Biotin)
Antibodies
Produced from sera of goats immunized with highly pure Recombinant Human Eotaxin (CCL11). Anti­Human Eotaxin (CCL11)­specific antibody was purified by affinity chromatography and then biotinylated.

Applications tested can vary by lot number. See Product Data Sheet for more information.

Type: Secondary
Antigen: Eotaxin (CCL11)
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Goat
Isotype: Goat IgG
Reactivity: Human
Order Now

SPECIFICATIONS

Antigen symbol Eotaxin (CCL11)
Environmentally Preferable
Antigen name Eotaxin
Antibody type Secondary
Clonality Polyclonal
Conjugation Biotin
Reactivity Human
Host Goat
Isotype IgG
Western blot Yes
ELISA Yes
Immunogen Recombinant Human Eotaxin (CCL11)
Form Lyophilized
Molecular weight 8.3 kDa
Purification Recombinant Human Eotaxin (CCL11)
Storage buffer See Product Data Sheet for more information.
Storage temperature Store at –20 °C prior to reconstitution.
Shipping temperature Ambient

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR