Anti-MCP-1 (CCL2) Mouse Monoclonal Antibody

Supplier: Peprotech
500-M71-500UG
10774-898EA 399.38 CAD
10774-898
Anti-MCP-1 (CCL2) Mouse Monoclonal Antibody
Antibodies
Monoclonal AntibodyProduced in BALB/c x ICR F1-mice immunized with highly pure Recombinant Human MCP-1 (CCL2). Anti­Human MCP-1 (CCL2)­specific antibody was purified from ascites fluid by antigen affinity chromatography.

Applications tested can vary by lot number. See Product Data Sheet for more information.

Type: Primary
Antigen: MCP-1 (CCL2)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: Mouse IgG1k
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR