Virus Recombinant MIP-2 (from E. coli)
Supplier: Peprotech
350-03-10UG 350-03-50UG 350-03-100UG 350-03-250UG 350-03-500UG 350-03-1MG
10774-410EA 209.84 CAD
10774-410 10774-412 10774-414 10774-416 10774-418 10774-420
Virus Recombinant MIP-2 (from E. coli)
Proteins and Peptides
Viral MIP-2 (vMIP-2) is a chemokine analog encoded by the human herpes virus, and has been shown to have antagonist activity towards several chemokine receptors. Recombinant Viral MIP-2 is a 7.9 kDa protein consisting of 70 amino acids, including the four highly conserved cysteine residues present in CC chemokines.
Ordering information: For research use only. Not for use in diagnostic or therapeutic procedures.