Human Eotaxin-3 (CCL26) Standard ABTS ELISA Development Kit, PeproTech, Inc.
Supplier: Peprotech
900-K167
10771-588KT 911.83 CAD
10771-588 75999-702
Human Eotaxin-3 (CCL26) Standard ABTS ELISA Development Kit, PeproTech, Inc.
Assays ELISAs
PeproTech's Human Eotaxin-3 (CCL26) ELISA Development Kit contains the key components required for the quantitative measurement of natural and/or recombinant human Eotaxin-3 (CCL26) in a sandwich ELISA format.
  • Easy-to-use
  • Optimized reagents for singleplex protein detection by solid phase sandwich enzyme-linked immunosorbent assay (ELISA)
Human Eotaxin-3 (CCL26) ELISA Development Kit contains the key components required for the quantitative measurement of natural and/or recombinant human Eotaxin-3 (CCL26) in a sandwich ELISA format. Using the ELISA protocol, the recommended microplates, reagents and solutions, the components supplied in this kit are sufficient to assay human Eotaxin-3 (CCL26) in approximately 1000 ELISA plate wells.