Murine IFN-γ Standard ABTS ELISA Development Kit, PeproTech
Supplier: Peprotech
900-K98
10771-548KT 911.83 CAD
10771-548 10777-848
Murine IFN-γ Standard ABTS ELISA Development Kit, PeproTech
Assays ELISAs
PeproTech's Murine IFN-γ ELISA development kit contains the key components required for the quantitative measurement of natural and/or recombinant murine IFN-γ in a sandwich ELISA format.
  • Easy-to-use
  • Optimized reagents for singleplex protein detection by solid phase sandwich enzyme-linked immunosorbent assay (ELISA)
Murine IFN-γ ELISA development kit contains the key components required for the quantitative measurement of natural and/or recombinant murine IFN-γ in a sandwich ELISA format. Using the ELISA protocol, the recommended microplates, reagents and solutions, the components supplied in this kit are sufficient to assay murine IFN-γ approximately 1000 ELISA plate wells.