VWR® Tissue Culture Plate Inserts, Polyester (PET) Membrane, Sterilized, Standard Line
Supplier: VWR International
10769-182 10769-184 10769-192 10769-194 10769-196 10769-204 10769-206 10769-214 10769-216 10769-218 10769-228 10769-238 10769-240 10769-242 76313-902 76313-904 76313-906
10769-182CS 284.08 CAD
10769-182 10769-184 10769-192 10769-194 10769-196 10769-204 10769-206 10769-214 10769-216 10769-218 10769-228 10769-238 10769-240 10769-242 76313-902 76313-904 76313-906
VWR® Tissue Culture Plate Inserts, Polyester (PET) Membrane, Sterilized, Standard Line
Microplates Insert Plates and Inserts
Tissue Culture Plate Inserts feature a thin, polyester membrane that is available in five pore sizes, ranging from 0.1 to 8.0 μm.
  • Temperature range: –20 to 50 °C (–4 to 122 °F)
  • Polyester membrane construction
  • Available in individual recyclable PET packaging
  • Inserts are not autoclavable
  • Non-pyrogenic
  • Transparent and translucent membrane options are available
Tissue Culture Plate Inserts provide excellent cell visibility under phase contrast microscopy and allow assessment of cell viability and monolayer formation. Tissue Culture Plate Inserts areMore Product Information