Spectra/Por® 1-5 Standard RC Dry Dialysis Trial Kits, Spectrum® Laboratories
Supplier: Spectrum Laboratories
132655T 132665T 132678T 132720T 132725T 132645T 132650T 132660T 132670T 132675T 132676T 132682T 132684T 132703T 132754T 132709T 132724T 132757T
97059-098EA 376.66 CAD
97059-098 97059-100 97059-102 97059-104 97059-106 97059-108 10768-526 10768-528 10768-530 10768-532 10768-534 10768-536 10768-538 10768-540 10768-542 10768-544 10768-546 10768-548 10768-550 10768-552 10768-554 10768-556 10768-558 10768-560 10768-562 10768-564 10768-566 10768-568 10768-570 10768-572 10768-574 10768-576 10768-642 10768-644 10768-646 10768-648 10768-650 10768-652
Spectra/Por® 1-5 Standard RC Dry Dialysis Trial Kits, Spectrum® Laboratories
Tubing Dialysis Tubing
Ideal for small volume dialysis or evaluation of membrane for an application, multi-purpose standard Regenerated Cellulose (RC) tubing is now available in 1 and 5 m length trial kits with a pair of tubing closures.
  • Hydrophilic, symmetric porosity
Standard Regenerated Cellulose (RC) is composed of natural cellulose regenerated from cotton linters, these membranes are perfectly suited for a broad range of laboratory dialysis. This clear and flexible membrane type with a symmetric porosity is economical and sturdy, ideal for desalting, buffer exchanges,More Product Information