Vertical Tube Compression Adapters, Ace Glass Incorporated
Supplier: Ace Glass
12731-60 12731-62 12731-64 12731-66 12731-68 12731-70 12731-72 12731-74 12731-30 12731-32 12731-34 12731-36 12731-38 12731-40 12731-42 12731-44 12731-46 12731-48 12731-50 12731-52 12731-02 12731-04 12731-06 12731-08 12731-10 12731-12 12731-14 12731-16 12731-18 12731-20 12731-22 12731-24 12731-26 12731-28 12731-54 12731-56 12731-58
10767-464EA 97.18 CAD
10767-464 10767-466 10767-468 10767-470 10767-472 10767-474 10767-476 10767-478 10767-812 10767-814 10767-816 10767-818 10767-820 10767-822 10767-824 10767-826 10767-828 10767-830 10767-832 10767-834 10767-980 10767-982 10767-984 10767-986 10767-988 10767-990 10767-992 10767-994 10767-996 10767-998 10768-000 10768-002 10768-004 10768-006 10768-008 10768-010 10768-012
Vertical Tube Compression Adapters, Ace Glass Incorporated
Adapters Adapters, Joints with Tube Connectors
These glass standard taper and spherical joint adapters feature a vertical ground glass sidearm.
  • Vertical sidearm
  • Glass [ST] or spherical joint
The sidearm is used to make tube compression fitting connections to a wide variety of flexible and rigid tubing using PTFE fittings.