Anti-CD5 Mouse Monoclonal Antibody (PerCP (Peridinin-Chlorophyll Protein Complex)-Cy5.5®) [clone: L17F12]

Supplier: Tonbo Biosciences
65-0058-T025 65-0058-T100 65-0058-T500
10753-796EA 237 CAD
10753-796 10753-798 10753-800
Anti-CD5 Mouse Monoclonal Antibody (PerCP (Peridinin-Chlorophyll Protein Complex)-Cy5.5®) [clone: L17F12]
Antibodies
The L17F12 antibody is specific for human CD5, a 67 kD transmembrane glycoprotein that is expressed on most thymocytes, mature T cells, and a subset of B cells. CD5 is a member of the scavenger receptor superfamily and is present on approximately 70% of normal peripheral blood lymphocytes. CD5 is involved in modulating antigen receptor signaling in both T and B cells and acts through binding of CD72, its receptor expressed on B cells.

Type: Primary
Antigen: CD5
Clonality: Monoclonal
Clone: L17F12
Conjugation: PerCP (Peridinin-Chlorophyll Protein Complex)-Cy5.5®
Epitope:
Host: Mouse
Isotype: IgG2a kappa
Reactivity:
Order Now

SPECIFICATIONS

Antigen symbol CD5
Antigen name CD5 molecule
Environmentally Preferable
Antigen synonyms CD5|LEU1|Lymphocyte antigen T1/Leu-1|T-cell surface glycoprotein CD5|T1
Antibody type Primary
Clonality Monoclonal
Conjugation PerCP (Peridinin-Chlorophyll Protein Complex)-Cy5.5®
Clone L17F12
Reactivity Human
Host Mouse
Gene ID 921
Isotype IgG2a kappa
Flow cytometry Yes
Cross adsorption No
Storage temperature Store undiluted at 2-8°C, protect from light, do not freeze.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR