Uncontrolled Electromantles, with Metal Case, Electrothermal
Supplier: Antyila Scientific
36103-04
10752-362EA 974.48 CAD
10752-362
Uncontrolled Electromantles, with Metal Case, Electrothermal
Heaters Heating Mantles
CMU heating mantles are used in wet chemistry to heat liquids in round bottom flasks
Ideal for use in fume hoods or remote working applications. The device must be used with a controller; recommended controllers are the MC5 and MC242. Electromantles accommodate round bottom flasks in capacities between 50mL and 5000mL. Element allows temperatures up to 450°C (842°F). Indicator lamps are provided for power and heater operation. Replaceable insulated heater cartridges are available. The rugged, easy to clean, powder-coated aluminium casing ensures long life and durability. Non-skid feet andMore Product Information