ASTM® Liquid-in-Glass Mercury Thermometers, Total Immersion, Nos. 7 to 67, Thermco
Supplier: Thermco
ACC007C ACC007F ACC008C ACC008F ACC012C ACC012F ACC013C ACC015C ACC015F ACC016C ACC016F ACC017C ACC017F ACC019F ACC020C ACC020F ACC021C ACC021F ACC022C ACC022F ACC026C ACC028C ACC028F ACC029C ACC030F ACC048F ACC050F ACC051F ACC054C ACC054F ACC056C ACC056F ACC058C ACC058F ACC060F ACC062C ACC062F ACC063C ACC063F ACC064C ACC064F ACC065C ACC065F ACC066C ACC066F ACC067C ACC067F ACC018C ACC018F ACC019C ACC044C ACC044F ACC045C ACC045F ACC046C ACC046F ACC047C ACC047F ACC048C ACC059C ACC059F ACC060C
10746-962EA 137.3 CAD
10746-962 10746-964 10746-966 10746-968 10746-982 10746-984 10746-986 10746-992 10746-994 10746-996 10746-998 10747-000 10747-002 10747-004 10747-006 10747-008 10747-118 10747-120 10747-122 10747-124 10747-132 10747-136 10747-138 10747-140 10747-142 10747-144 10747-166 10747-170 10747-172 10747-176 10747-180 10747-182 10747-184 10747-186 10747-192 10747-194 10747-196 10747-202 10747-204 10747-206 10747-208 10747-210 10747-212 10747-214 10747-216 10747-218 10747-220 10747-222 10747-224 10747-760 10747-762 10747-764 10747-778 10747-780 10747-782 10747-784 10747-786 10747-788 10747-790 10747-792 10747-794 10747-796 10747-798 10747-800
ASTM® Liquid-in-Glass Mercury Thermometers, Total Immersion, Nos. 7 to 67, Thermco
Thermometers Analog Thermometers
These high precision liquid-in-glass mercury thermometers are hand blown, which bear graduations, numbers and letters etched into the glass and filled with and inert gas above the mercury column
ASTM thermometers are used for a variety of specified tests which quantify and qualify repeatable test results in the fields of oil and petroleum testing.
These thermometers were designed for specific testing and can be calibrated to your specific requirements.
Ordering information: Accessories not included. Certificate of calibration not included. ASTM thermometers with a NIST TraceableMore Product Information