Peristaltic Pumps, rotarus®, HIRSCHMANN
Supplier: Hirschmann
9501322 9501324 9501342 9501344 9501362 9501364 9501522 9501524 9501542 9501544 9501562 9501564
10713-846EA 8847.75 CAD
10713-846 10713-848 10713-850 10713-852 10713-854 10713-856 10713-858 10713-860 10713-862 10713-864 10713-866 10713-876
Peristaltic Pumps, rotarus®, HIRSCHMANN
Pumps Peristaltic Pumps
The rotarus® peristaltic pump series with a selection of different motors, varying housing safety classes and intelligent control of delivery volumes, covering a broad spectrum of application areas in the lab or industry. 50 and 100 W motors ensure precise delivery in speed ranges from 0.2 to 500 min⁻¹. This means that media with a high viscosity can also be accurately dispensed. Configuration data for basic parameters can also be stored in this manner an retrieved at any time. The flow and volume models have automatic blockage detection an tube rupture monitoring.
  • Changeable pump
More Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable
IP rating IP54 IP65