Reusable Plastic Animal Cages, Tecniplast®
Supplier: Tecniplast Usa
1144B00SU 1144B001 1144B00139 1144B00139 1264C001 1354G001 2000P00130 2150E001 2150E00SU 1264C00139 1264C00SU
10710-021CS 3289.42 CAD
10710-021 10710-022 10712-110 10712-234 10712-318 10712-320 77322-362 77322-370 77322-398 77322-406 77322-356 77322-416 77322-372 77322-374
Reusable Plastic Animal Cages, Tecniplast®
Animal Supplies Animal Cages
Plastic animal cages provide warm surfaces ideal for housing laboratory rodents and are convenient to use and maintain. Cage bodies are designed with corner lugs (side-arms) for stackable storage without scratching or jamming. Autoclavable cage bodies are available:
  • Clear standard polycarbonate
  • Clear polycarbonate with grommet for access to sipper tube on rack
  • Clear polycarbonate with cut-away bottoms for removing waste products
  • High-temperature clear polysulfone for repeated autoclaving and better chemical resistance
  • polyetherimide for maintaining
More Product Information