VWR® Buchner Funnels with Fritted Disc
Supplier: VWR International
256.M.01 256.F.01 256.C.02 256.M.02 256.F.03 256.C.04 256.M.04 256.F.04 256.C.05 256.M.05 256.F.05 256.C.06 256.M.06 256.F.06 256.C.07 256.M.07 256.F.07 256.C.08 256.M.08 256.C.09 256.M.09 256.F.02 256.C.03 256.M.03
10545-868CS 414.15 CAD
10545-868 10545-870 10545-872 10545-874 10545-876 10545-878 10545-880 10545-882 10545-884 10545-886 10545-888 10545-890 10545-892 10545-894 10545-896 10545-898 10545-900 10545-902 10545-904 10545-906 10545-908 10546-040 10546-042 10546-044
VWR® Buchner Funnels with Fritted Disc
Funnels Filtration Funnels Büchner Funnels
Ideal for use where filter paper or membranes cannot withstand chemical attack of the filtrate.
  • Borosilicate glass fabrication
  • Fritted disc
  • Beaded top rims
Funnels feature medium-walled stems and beaded top rims for extra strength.
Made from ASTM E438 Type 1 Class A borosilicate 33 expansion glass. Funnel meets federal specification DD- F -776.
These funnels are used in qualitative inorganic analysis and preparative chemistry and are resistant to corrosion and heat.
Order Now
SPECIFICATIONS
Environmentally Preferable