VWR® Erlenmeyer Flasks, Narrow Mouth
Supplier: VWR International
10536-676 10536-678 10536-912 10536-914 10536-924 10536-926 10545-842 10545-844 10545-846 10545-848 75870-512
10536-676CS 553.78 CAD
10536-676 10536-678 10536-912 10536-914 10536-924 10536-926 10545-842 10545-844 10545-846 10545-848 75870-512
VWR® Erlenmeyer Flasks, Narrow Mouth
Flasks Erlenmeyer Flasks
Flasks are durable and come with reinforced rim for easy pouring and also avoids any breakage while washing.
  • Borosilicate glass fabrication
  • Durable design
Feature durable white ceramic enamel scale which indicate approximate volume for measuring and mixing.
Manufactured from ASTM E438 Type 1 Class A borosilicate 33 expansion glass. As per ASTM specification E 1404 Type 1.
Order Now
SPECIFICATIONS
Environmentally Preferable