Anti-APPL1 Polyclonal Antibody Pair

Supplier: Abnova
H00026060-PW1
10536-290EA 1032.31 CAD
10536-290
Anti-APPL1 Polyclonal Antibody Pair
Antibodies
This Immunopreciptitation/ Western Blot Antibody Pair set comes with one Antibody for Immunoprecipitation and another to detect the precipitated protein in Western Blot.

Antibody pair set content: 1. Antibody pair for IP: Rabbit Polyclonal Anti-APPL1 (300 µl); 2. Antibody pair for Western Blot: Mouse Polyclonal Anti-APPL1 (50 µl)

Type:
Antigen: APPL1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Multiple
Isotype:
Reactivity: Human
Order Now

SPECIFICATIONS

Antigen symbol APPL1
Antigen name Adaptor Protein, Phosphotyrosine Interaction, PH Domain And leucine zipper Containing 1
Environmentally Preferable
Antigen synonyms XAP4|RPB5|hRPB25|hsRPB5|RPABC1
Clonality Polyclonal
Reactivity Human
Host Multiple
Gene ID 26060
Isotype
Western blot Yes
ImmunoPrecipitation Yes

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR