Anti-ACP6 Mouse Polyclonal Antibody

Supplier: Abnova
H00051205-B01P
10510-176EA 529.3 CAD
10510-176
Anti-ACP6 Mouse Polyclonal Antibody
Antibodies
Type: Primary
Antigen: ACP6
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

SPECIFICATIONS

Antigen symbol ACP6
Antigen name Acid Phosphatase 6, lysophosphatidic
Environmentally Preferable
Antigen synonyms LPAP|PACPL1|ACPL1
Antibody type Primary
Clonality Polyclonal
Conjugation Unconjugated
Reactivity Human
Host Mouse
Gene ID 51205
Isotype
Western blot Yes
Cross adsorption No
Storage temperature Cool
Shipping temperature Ice

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR