Anti-MRAS Mouse Polyclonal Antibody

Supplier: Abnova
H00022808-B01P
10509-712EA 529.3 CAD
10509-712
Anti-MRAS Mouse Polyclonal Antibody
Antibodies
Type: Primary
Antigen: MRAS
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

SPECIFICATIONS

Antigen symbol MRAS
Environmentally Preferable
Antigen name Muscle RAS Oncogene homolog
Antigen synonyms M-RAs|RRAS3|R-RAS3
Antibody type Primary
Clonality Polyclonal
Conjugation Unconjugated
Reactivity Human
Host Mouse
Gene ID 22808
Isotype
Western blot Yes
Cross adsorption No
Storage temperature Cool
Shipping temperature Ice

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR