GsBP®-InoWax Polar, PEG GC Columns, GS-Tek
Supplier: General Separation Technologies, Inc.
2032-6001 2032-6002 2032-6005 2032-6010 2018-1001 2018-2001 2018-2003 2018-4001 2018-4003 2020-2502 2020-2504 2020-3002 2025-0501 2025-1501 2025-1502 2025-1505 2025-3001 2025-3002 2025-3005 2025-6001 2025-6002 2025-6005 2032-1502 2032-1505 2032-1510 2032-3001 2032-3002 2032-3005 2032-3010 2032-5010 2053-1505 2053-1510 2053-1520 2053-2520 2053-3002 2053-3005 2053-3010 2053-3015 2053-3020 2053-6010 2025-3002-6PK 2032-3002-6PK
10499-348EA 2785.87 CAD
10499-348 10499-350 10499-352 10499-354 10499-430 10499-432 10499-434 10499-436 10499-438 10499-440 10499-442 10499-444 10499-452 10499-454 10499-456 10499-458 10499-460 10499-462 10499-466 10499-468 10499-470 10499-472 10499-474 10499-476 10499-478 10499-480 10499-482 10499-486 10499-488 10499-490 10499-492 10499-494 10499-496 10499-498 10499-508 10499-510 10499-512 10499-514 10499-516 10499-518 10510-892 10510-894
GsBP®-InoWax Polar, PEG GC Columns, GS-Tek
Chromatography Columns
Typical application: Alcohols II and III, Amines (low MW), Amines/ Alcohols/ Chlorides, Aromatics analysis: ASTM D16 analytes, Aromatics II, Blood alcohol, Bourbon, Branched alcohols, BTEX, Chlorinated Isooctane, Citronella java oil, Esters III, Ethylene oxide, FAME Standard I, FAMEs (Coca Butter), Fast styrene analysis, Flavor volatiles, Formaldehyde Underivatized, Fragrance allergens, Fragrance Reference Standard II, Free Fatty Acids, Free Organic Acids/C4-C5 Isomers, Impurities in Ethylbenzene, Impurities in mixed xylenes, Impurities in p-Xylene-ASTM D3798, Impurities in styrene,More Product Information