GsBP®-5MS Non-Polar GC Columns, GS-Tek
Supplier: General Separation Technologies, Inc.
1518-2001 1518-2001-625 1518-2001PAH 1518-2001UI 1518-2003 1518-2003-625 1518-2003UI 1518-4001 1520-1203 1520-1703 1520-2503 1520-5003 1525-1501 1525-1502 1525-1502UI 1525-1505 1525-1510 1525-1510UI 1525-2502 1525-2502UI 1525-3001 1525-3002 1525-3002-625 1525-3002PAH 1525-3002UI 1525-3005 1525-3005-625 1525-3005UI 1525-3010 1525-3010-625 1525-3010UI 1525-5002 1525-5002UI 1525-6001 1525-6002 1525-6002-625 1525-6002PAH 1525-6002UI 1525-6005 1525-6010 1525-6010UI 1532-1501 1532-1502 1532-1502PAH 1532-1505 1532-1510 1532-2505 1532-3001 1532-6001 1532-6002 1532-6002UI 1532-3002 1532-3002-625 1532-3002UI 1532-3005 1532-3005-625 1532-3005UI 1532-3010 1532-3010UI 1532-6005 1532-6010 1532-6010UI 1553-1515 1553-3005 1553-3010 1553-3015 1525-3002-6PK
10499-138EA 1365.58 CAD
10499-138 10499-140 10499-142 10499-144 10499-146 10499-148 10499-150 10499-152 10499-154 10499-156 10499-158 10499-160 10499-162 10499-164 10499-166 10499-168 10499-170 10499-172 10499-174 10499-176 10499-178 10499-180 10499-182 10499-184 10499-186 10499-188 10499-190 10499-192 10499-194 10499-196 10499-198 10499-200 10499-204 10499-206 10499-208 10499-210 10499-212 10499-262 10499-264 10499-266 10499-268 10499-270 10499-272 10499-274 10499-276 10499-278 10499-280 10499-282 10499-284 10499-286 10499-288 10499-290 10499-332 10499-334 10499-338 10499-340 10499-342 10499-344 10499-346 10499-356 10499-358 10499-360 10499-362 10499-364 10499-366 10499-368 10499-370 10499-372 10499-374 10499-376 10499-378 10499-380 10499-382 10499-384 10499-386 10510-888
GsBP®-5MS Non-Polar GC Columns, GS-Tek
Chromatography Columns
Typical Application: Acidic/neutral drugs, alkaloids, amines and nitriles, amphetamine and methamphetamine, antiepileptic, basic drugs, chlorinated pesticides, EPA method 508, CLP standard, semivolatile organics, diesel fuel, drug of abuse, endocrine disruptors: alkyl phenols, endocrine disruptors: phthalate, EPA 608.1, EPA air analysis method TO-15, EPA method 525.2, EPA method 551.1, EPA method 610, EPA method 8061 (phthalate ester), EPA method 8270, EPA-625 phenols, flavor mixture, food packaging volatiles, formaldehyde, 50ppb, fragrance allergens, gasoline, halogenated compounds, localMore Product Information