Acid Reagent Powder Pillows for Chromium, Hach
Supplier: Hach
1421199 1432399 1436369 1432298 2081599 87299 2380268 2244100
CA11017-092PK 167.84 CAD
CA11017-092 104085-386 104085-422 104085-426 104085-420 104085-744 104086-988 104086-178 104088-188
Acid Reagent Powder Pillows for Chromium, Hach
Environmental Test Kits
For total chromium determination by the alkaline hypobromite oxidation method.
Order Now
SPECIFICATIONS
Environmentally Preferable
Method Alkaline Hypobromite Oxidation