Human Recombinant GUCY2C (from HEK293 cells)

Supplier: ACROBIOSYSTEMS INC MS

GUCH52H5100UG
103871-718EA 568.8 CAD
103871-718
Human Recombinant GUCY2C (from HEK293 cells)
Proteins and Peptides

Human GUCY2C / Guanylyl cyclase C Protein, His Tag, ACROBiosystems

Order Now

SPECIFICATIONS

Protein/peptide name GUCY2C
Environmentally Preferable
Protein synonyms GUCY2C|hSTAR|STA receptor|GC-C|STAR|GUC2C
Protein/peptide type Recombinant
Species Human
Source HEK293 cells
Conjugation Unconjugated
Gene ID P25092-1
Endotoxin level Low
Endotoxin content Less than 1.0 EU per μg by the LAL method.
Biological activity 2e5 of anti-GUCY2C CAR-293 cells were stained with 100 µL of 1 µg/mL Human GUCY2C, His Tag (Cat. No. GUC-H52H5) and negative control protein respectively, washed and then followed by PE anti-His antibody and analyzed with FACS (Routinely tested).
Purity >90% as determined by SDS-PAGE.
Formulation Lyophilized from 0.22 μm filtered solution in PBS, pH7.4. Normally trehalose is added as protectant before lyophilization.
Molecular weight 47.9 kDa
Storage conditions –20 °C
Shipping temperature RT
Tag sequence His Tag

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR