Bevacizumab ELISA Assay Kit, Eagle Biosciences, Inc.
Supplier: Eagle Biosciences
IG-AA104
103389-212EA 2818.46 CAD
103389-212
Bevacizumab ELISA Assay Kit, Eagle Biosciences, Inc.
Assays ELISAs
Bevacizumab ELISA analyzes biologically active form of free bevacizumab in serum and plasma samples.
  • Dynamic Range: 6 -200 ng/mL
  • Sensitivity: 2 ng/mL
  • Incubation: 2 hours
The Eagle Biosciences Bevacizumab (Avastin®) ELISA Assay Kit is an enzyme immunoassay for the precise analytical determination of free Bevacizumab in serum and plasma samples.  The Bevacizumab (Avastin®) ELISA Assay Kit is for research use only and not to be used in diagnostic procedures. 