FSH ELISA, Eagle Biosciences, Inc.
Supplier: Eagle Biosciences
DCM010
103389-066EA 550.77 CAD
103389-066
FSH ELISA, Eagle Biosciences, Inc.
Assays ELISAs
FSH EIA is direct solid phase assay to detect Follicle-Stimulating Hormone in human serum or plasma.
  • Dynamic Range: 5 -100 mIU/ml
  • Sensitivity: 0.17 mIU/ml
  • Incubation: 1.5 hours
Follicle Stimulating hormone (FSH) is a glycoprotein consisting of two subunits with an approximate molecular mass of 35,500 daltons. The α- subunit is similar to other pituitary hormones [luteinizing stimulating hormone (LH), thyroid stimulating hormone (TSH) and chorionic gonadotropin (hCG)] while the β-subunit is unique. The β- subunit confers the biological activity to the molecule. Stimulation by gonadotropin-releasing hormone (GnRH) causes release of FSH, as well as LH, from the pituitary and is transported by the blood to their sites of action, the testes or ovary.
In men, FSH acts on the Sertoli cells of the testis, stimulating the synthesis of inhibin, which appears to specifically inhibit further FSH secretion, and androgen-binding protein. Thus, it indirectly supports spermatogenesis.
In women, FSH acts on the granulosa cells of the ovary, stimulatin steroidogensis. All ovulatory menstrual cycles have a characteristic pattern of FSH, as well as LH, secretion. The menstrual cycle is divided into a follicular phase and a luteal phase by the midcycle surge of the gonadotropins (LH and FSH). As the follicular phase progresses, FSH concentration decreases. Near the time ovulation occur, about midcycle, FSH peaks (lesser in magnitude than LH) to its highest level.
The clinical usefulness of the measurement of Follicle Stimulating hormone (FSH) in ascertaining the homeostasis of fertility regulation via the hypothalamic - pituitary - gonadal axis has been well established (1,2).