Anti-PMS2 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 163C1251]

Supplier: Novus Biologicals
NB100-56554PE
103360-172EA 776.92 CAD
103360-172
Anti-PMS2 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 163C1251]
Antibodies
The PMS2 Antibody (163C1251) [PE] from Novus Biologicals is a mouse monoclonal antibody to PMS2. This antibody reacts with human, mouse. The PMS2 Antibody (163C1251) [PE] has been validated for the following applications: Flow Cytometry.

Type: Primary
Antigen: PMS2
Clonality: Monoclonal
Clone: 163C1251
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR