G57 Pharmaceutical PSA Patterns, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
68091-20.4 68091-23 68091-24.5 68091-25 68091-27
103306-218EA 781.28 CAD
103306-218 103306-220 103306-222 103306-224 103306-228
G57 Pharmaceutical PSA Patterns, Electron Microscopy Sciences
Graticules, Reticles and Grids
This graticule was designed for the pharmaceutical industry.
  • Scale is incorporated
  • Glass sandwich surface
However, it is also useful where particle size considerations are restricted to 10 and 25 mm. Dots and circles give quick references for these two sizes.
The microscope must be calibrated when ordering this graticule, such that the circle must equate to one mm on the microscope stage.