Formvar Support Film on Square Grids, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
FF100-NI-25 FF200-NI-25 FF400-CU-50 FF400-CU-25 FF200-CU-50 FF150-CU-50 FF100-CU-50 FF150-NI-50 FF300-AU-25 FF100-CU-UL FF150-CU-UL FF200-CU-UL FF300-CU-UL FF400-CU-UL FF100-CU-TH FF150-CU-TH FF200-CU-TH FF300-CU-TH FF400-CU-TH FF100-CU-ET FF150-CU-ET FF200-CU-ET FF300-CU-ET FF400-CU-ET FF100-NI-UL FF150-NI-UL FF200-NI-UL FF300-NI-UL FF400-NI-UL FF100-NI-TH FF150-NI-TH FF200-NI-TH FF300-NI-TH FF400-NI-TH FF100-NI-ET FF150-NI-ET FF200-NI-ET FF300-NI-ET FF400-NI-ET FF100-AU-UL FF150-AU-UL FF200-AU-UL FF300-AU-UL FF400-AU-UL FF100-AU-TH FF150-AU-TH FF200-AU-TH FF300-AU-TH FF400-AU-TH FF100-AU-ET FF150-AU-ET FF200-AU-ET FF300-AU-ET FF400-AU-ET
100491-138EA 127.74 CAD
100491-138 100491-158 100491-176 100491-174 100491-156 100491-146 100491-136 100491-150 100491-162 103302-172 103302-174 103302-176 103302-178 103302-180 103302-196 103302-198 103302-200 103302-202 103302-204 103302-228 103302-230 103302-232 103302-234 103302-236 103302-794 103302-796 103302-798 103303-064 103303-066 103303-196 103303-198 103303-200 103303-202 103303-204 103303-220 103303-222 103303-224 103303-226 103303-228 103303-324 103303-326 103303-328 103303-330 103303-332 103303-388 103303-390 103303-392 103303-394 103303-396 103303-486 103303-488 103303-490 103303-492 103303-494
Formvar Support Film on Square Grids, Electron Microscopy Sciences
Graticules, Reticles and Grids
A thin film of pure formvar resin coats the shiny side of a square grid.
  • Formvar coatings thickness of standard (10 nm), ultra-thin (5 to 6 nm), thick (15 to 20 nm), or extra thick (25 to 50 nm)
  • Select from copper, nickel, and gold grids in 100, 150, 200, 300, and 400 mesh sizes
  • Optically checked followed by batch testing in the EM
Film is laid on the shiny side of the grid.
Packaging: Packed in grid storage box.