Formvar Carbon Support Film on Hexagonal Grids, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
FCF100H-CU-25 FCF100H-CU-50 FCF200H-CU-25 FCF200H-CU-50 FCF200H-NI-50 FCF300H-CU-25 FCF300H-CU-50 FCF300H-NI-25 FCF400H-CU-25 FCF400H-CU-50 FCF400H-NI-25 FCF200H-CU-SB FCF200H-CU-SC FCF200H-CU-UA FCF200H-CU-UB FCF200H-CU-UC FCF200H-CU-TA FCF200H-CU-TB FCF200H-CU-TC FCF200H-CU-EA FCF200H-CU-EB FCF200H-CU-EC FCF300H-CU-SB FCF300H-CU-SC FCF300H-CU-UA FCF300H-CU-UB FCF300H-CU-UC FCF300H-CU-TA FCF300H-CU-TB FCF300H-CU-TC FCF300H-CU-EA FCF300H-CU-EB FCF300H-CU-EC FCF400H-CU-SB FCF400H-CU-SC FCF400H-CU-UA FCF400H-CU-UB FCF400H-CU-UC FCF400H-CU-TA FCF400H-CU-TB FCF400H-CU-TC FCF400H-CU-EA FCF400H-CU-EB FCF400H-CU-EC FCF300H-NI-SB FCF300H-NI-SC FCF300H-NI-UA FCF300H-NI-UB FCF300H-NI-UC FCF300H-NI-TA FCF300H-NI-TB FCF300H-NI-TC FCF300H-NI-EA FCF300H-NI-EB FCF300H-NI-EC FCF400H-NI-SB FCF400H-NI-SC FCF400H-NI-UA FCF400H-NI-UB FCF400H-NI-UC FCF400H-NI-TA FCF400H-NI-TB FCF400H-NI-TC FCF400H-NI-EA FCF400H-NI-EB FCF400H-NI-EC FCF100H-CU-SB FCF100H-CU-UA FCF100H-NI-EC FCF200H-NI-EA FCF100H-NI-SC FCF100H-NI-UA FCF200H-NI-SB FCF200H-NI-SC FCF200H-NI-UA FCF200H-NI-UB FCF200H-NI-UC FCF100H-CU-EB FCF100H-CU-EC FCF100H-NI-TB FCF100H-NI-TC FCF200H-NI-TA FCF200H-NI-TB FCF200H-NI-TC FCF200H-NI-EB FCF200H-NI-EC FCF200H-AU-UB FCF200H-AU-UC FCF300H-AU-SB FCF300H-AU-SC FCF200H-AU-SB FCF200H-AU-SC FCF200H-AU-UA FCF300H-AU-UA FCF300H-AU-UB FCF300H-AU-UC FCF400H-AU-SB FCF400H-AU-SC FCF400H-AU-UA FCF400H-AU-UB FCF400H-AU-UC FCF200H-AU-TA FCF200H-AU-TB FCF200H-AU-TC FCF300H-AU-TA FCF300H-AU-TB FCF300H-AU-TC FCF400H-AU-TA FCF400H-AU-TB FCF400H-AU-TC FCF200H-AU-EA FCF200H-AU-EB FCF200H-AU-EC FCF300H-AU-EA FCF300H-AU-EB FCF300H-AU-EC FCF400H-AU-EA FCF400H-AU-EB FCF400H-AU-EC FCF100H-AU-UB FCF100H-AU-TB FCF100H-AU-EC
102100-470BX 112.86 CAD
102100-470 102100-472 102100-506 102100-508 102100-512 102100-530 102100-532 102100-534 102100-542 102100-544 102100-546 103302-438 103302-440 103302-442 103302-444 103302-702 103302-704 103302-706 103302-708 103302-852 103302-854 103302-856 103302-918 103302-920 103302-922 103302-924 103302-926 103302-928 103302-930 103302-932 103302-940 103302-942 103302-944 103303-008 103303-010 103303-012 103303-014 103303-016 103303-018 103303-020 103303-022 103303-024 103303-026 103303-028 103303-150 103303-152 103303-154 103303-156 103303-158 103303-160 103303-162 103303-164 103303-166 103303-168 103303-170 103303-172 103303-174 103303-176 103303-178 103303-180 103303-182 103303-184 103303-186 103303-188 103303-190 103303-192 103303-258 103303-262 103303-286 103303-288 103303-296 103303-298 103303-304 103303-306 103303-308 103303-310 103303-312 103303-344 103303-346 103303-378 103303-380 103303-382 103303-384 103303-386 103303-450 103303-452 103304-516 103304-518 103304-520 103304-522 103304-678 103304-680 103304-682 103304-684 103304-686 103304-688 103304-690 103304-692 103304-694 103304-696 103304-698 103304-940 103304-942 103304-944 103304-946 103304-948 103304-950 103304-952 103304-954 103304-956 103304-996 103304-998 103305-000 103305-002 103305-004 103305-006 103305-008 103305-010 103305-012 103305-052 103305-086 103305-136
Formvar Carbon Support Film on Hexagonal Grids, Electron Microscopy Sciences
Graticules, Reticles and Grids
The formvar coated grids are stabilized with an evaporated carbon film. This type of coating is excellent for specimen support, especially for ultra thin sections. All film is laid on the shiny side of the grid.
  • Formvar coatings thickness of standard (10 nm), ultra-thin (5 to 6 nm), thick (15 to 20 nm), or extra thick (25 to 50 nm)
  • Select copper, nickel, or gold grids in 100, 150, 200, 300 or 400 mesh
  • Carbon coatings thickness of 1 nm, 3 to 4 nm, and 20 to 30 nm
  • Optically checked followed by batch testing in the EM
Packaging: Packed in grid storage box.