Formvar Carbon Support Film on Square Grids, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
FCF200-CU FCF400-NI-50 FCF100-NI-25 FCF300-CU-50 FCF400-NI-25 FCF100-CU-50 FCF200-AU-50 FCF200-NI-50 FCF400-CU FCF150-CU-50 FCF200-NI-25 FCF300-NI-25 FCF400-AU-50 FCF200-CU-50 FCF300-CU FCF100-NI-50 FCF200-AU-25 FCF400-AU-25 FCF100-AU-50 FCF300-NI-50 FCF100-AU-25 FCF300-AU-25 FCF150-AU-50 FCF150-CU-25 FCF150-NI-50 FCF400-CU-50 FCF150-CU-TA FCF150-CU-TB FCF100-CU-SB FCF100-CU-SC FCF100-CU-UA FCF100-CU-UB FCF100-CU-UC FCF200-CU-UA FCF200-CU-UB FCF200-CU-UC FCF200-CU-TA FCF200-CU-TB FCF200-CU-TC FCF150-CU-EA FCF150-CU-EB FCF200-CU-SB FCF200-CU-SC FCF300-CU-SB FCF300-CU-SC FCF300-CU-UA FCF300-CU-UB FCF300-AU-UA FCF300-AU-UB FCF300-AU-UC FCF400-AU-SB FCF100-CU-TA FCF100-CU-TB FCF100-CU-EA FCF100-CU-EB FCF100-CU-EC FCF150-CU-SB FCF150-CU-SC FCF150-CU-UA FCF200-CU-EA FCF200-CU-EB FCF200-CU-EC FCF300-CU-UC FCF400-CU-SB FCF400-CU-SC FCF400-CU-UA FCF200-NI-TA FCF200-NI-TB FCF400-CU-UB FCF400-CU-UC FCF300-CU-TA FCF300-CU-TB FCF300-CU-TC FCF400-CU-TA FCF400-CU-TB FCF400-CU-TC FCF100-NI-SB FCF100-NI-UA FCF100-NI-UB FCF200-NI-SB FCF200-NI-SC FCF200-NI-UA FCF200-NI-UB FCF200-NI-UC FCF300-CU-EA FCF300-CU-EB FCF300-CU-EC FCF300-NI-SB FCF300-NI-SC FCF300-NI-UA FCF300-NI-UB FCF300-NI-UC FCF400-CU-EA FCF400-CU-EB FCF400-CU-EC FCF400-NI-SB FCF400-NI-SC FCF400-NI-UA FCF400-NI-UB FCF400-NI-UC FCF100-NI-TA FCF100-NI-TB FCF150-NI-SB FCF150-NI-SC FCF150-NI-UA FCF200-NI-TC FCF300-NI-TA FCF300-NI-TB FCF300-NI-TC FCF400-NI-TA FCF400-NI-TB FCF400-NI-TC FCF150-NI-TA FCF100-NI-EA FCF100-NI-EB FCF200-NI-EA FCF200-NI-EB FCF200-NI-EC FCF300-NI-EA FCF300-NI-EB FCF300-NI-EC FCF400-NI-EA FCF400-NI-EB FCF400-NI-EC FCF400-AU-SC FCF400-AU-UA FCF400-AU-UB FCF400-AU-UC FCF150-AU-SB FCF200-AU-SB FCF200-AU-SC FCF200-AU-UA FCF200-AU-UB FCF200-AU-UC FCF300-AU-SB FCF300-AU-SC FCF150-AU-TB FCF200-AU-TA FCF200-AU-TB FCF200-AU-TC FCF300-AU-TA FCF300-AU-TB FCF300-AU-TC FCF400-AU-TA FCF400-AU-TB FCF400-AU-TC FCF150-AU-EC FCF200-AU-EA FCF200-AU-EB FCF200-AU-EC FCF300-AU-EA FCF300-AU-EB FCF300-AU-EC FCF400-AU-EA FCF400-AU-EB FCF400-AU-EC FCF100-AU-SB FCF100-AU-UA FCF100-AU-EC FCF100-AU-TB FCF100-AU-TC
100491-092EA 124.55 CAD
100491-092 100491-130 100491-076 100491-116 100491-128 100491-074 100491-090 100491-098 100491-124 100491-084 100491-096 100491-118 100491-122 100491-094 100491-114 100491-078 100502-056 100502-060 100491-070 100491-086 100491-120 100502-052 100502-054 100502-058 100491-080 100491-082 100491-088 100491-126 103302-246 103302-248 103302-264 103302-266 103302-268 103302-270 103302-272 103302-274 103302-276 103302-278 103302-280 103302-282 103302-284 103302-428 103302-430 103302-434 103302-436 103302-520 103302-522 103302-524 103302-526 103302-726 103302-728 103302-730 103302-732 103302-756 103302-758 103302-762 103302-764 103302-766 103302-768 103302-770 103302-772 103302-778 103302-780 103302-782 103302-784 103302-786 103302-788 103302-790 103302-804 103302-806 103302-848 103302-850 103302-906 103302-908 103302-910 103302-912 103302-914 103302-916 103302-950 103302-954 103302-956 103302-960 103302-962 103302-964 103302-966 103302-968 103302-970 103302-972 103302-974 103302-976 103302-978 103302-980 103302-982 103302-984 103302-986 103302-988 103302-990 103302-992 103302-994 103302-996 103302-998 103303-000 103303-068 103303-070 103303-074 103303-076 103303-078 103303-080 103303-082 103303-084 103303-086 103303-088 103303-090 103303-092 103303-106 103303-120 103303-122 103303-132 103303-134 103303-136 103303-138 103303-140 103303-142 103303-144 103303-146 103303-148 103303-274 103303-276 103303-278 103303-280 103303-426 103303-436 103303-438 103303-440 103303-442 103303-444 103303-446 103303-448 103303-556 103303-560 103303-562 103303-564 103303-566 103303-568 103303-570 103303-572 103303-574 103303-576 103303-646 103303-648 103303-650 103303-652 103303-654 103303-656 103303-658 103303-660 103303-662 103303-664 103303-898 103303-902 103303-912 103304-090 103304-092
Formvar Carbon Support Film on Square Grids, Electron Microscopy Sciences
Graticules, Reticles and Grids
The formvar coated grids are stabilized with an evaporated carbon film.
  • Formvar coatings thickness of standard (10 nm), ultra-thin (5 to 6 nm), thick (15 to 20 nm), or extra thick (25 to 50 nm)
  • Carbon coatings thickness of 1 nm, 3 to 4 nm, and 20 to 30 nm
  • Select copper, nickel, or gold grids in 100, 150, 200, 300, or 400 mesh
  • Optically checked followed by batch testing in the EM
This type of coating is excellent for specimen support, especially for ultra thin sections. All film is laid on the shiny side of the grid.
Packaging: Packed in grid storage box.