Ergonomic ESD Tweezers Style 5, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
03045SAESDET1 0304-5SAESD-ET2
103302-606EA 94.29 CAD
103302-606 103302-608
Ergonomic ESD Tweezers Style 5, Electron Microscopy Sciences
Forceps
These precision tweezers from Dumont made of high quality steels with perfect finish suits all application needs.
The anti-magnetic steel ergonomic tweezers have a soft electro static discharge (ESD) grip. The red rubber handle has very high resistivity and a comfortable grip. Antimagnetic (80%) contains C, Cr, Ni and Mo and has good resistance to corrosion due to a high concentration of molybdenum but is not as hard as Inox.
Length is 110 mm.