Synaptek® Beryllium-Copper Grids, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
S2010-NN S2010-ND SG2010-NN S2010-NUM S2010-DOT S2010-NOTCH S1020-NI S1500-DOT S1500NI-DOT S1500MO-DOT S1808-ND
100496-012EA 204.04 CAD
100496-012 100496-010 100496-020 100496-016 102101-522 102101-524 103301-812 103301-814 103301-816 103302-142 103302-540
Synaptek® Beryllium-Copper Grids, Electron Microscopy Sciences
Graticules, Reticles and Grids
Unflexible grids, made of a special beryllium-copper alloy, nickel and molybdenum.
  • Extreme stability for coating with support film
  • Contamination free and reusable after cleaning
  • Diameter 3.05 mm
  • Very reliable under the electron beam
NUM grids: Numbered grids are in random order. Numbers may be duplicated.
DOT grids: 2 dots are marked on one side of the grid for identification. Dots are visible to the naked eye.
NOTCH grids: A mark, stamped on one side of the grid to facilitate the handling of the grids. Notch is available with NUM or DOT grids.
More Product Information