Mouse Recombinant Angiopoietin-related protein 4
Supplier: Bon Opus Biosciences
C761-10UG C761-50UG C761-500UG C761-1MG
102942-730EA 158 CAD
102942-730 102942-732 102942-734 102942-736
Mouse Recombinant Angiopoietin-related protein 4
Proteins and Peptides
Bon Opus Biosciences offers quality recombinant proteins products covering a broad collection of cytokines, enzymes, diagnostic and detection reagents, and other protein-related products
  • Consistent Sizing options
  • Datasheet Included
  • High Purity, Low Endotoxin levels
Recombinant Mouse Angiopoietin Like protein 4 is produced by our Mammalian expression system and the target gene encoding Lys167-Ser410 is expressed with a Fc tag at the C-terminus.
Accessories information: Accessories not included.
Delivery information: Shipped Via FedEx
Packaging: Shipped domestic on polar packs in a 9"x6"x7" box, or shipped internationally on Dry Ice in a 14" cube box.
Caution: This product list contains purified protein and other non-biological reagents, which are minimum risk biologicals, non-toxic and non-hazardous, and are intended for laboratory research use only.
The products are not intended for human or diagnostic use, and shall not be injected or ingested by any human.