Anti-TEX19 Rat Monoclonal Antibody [clone: 678826]

Supplier: R&D Systems
MAB66451
102886-582EA 833.85 CAD
102886-582
Anti-TEX19 Rat Monoclonal Antibody [clone: 678826]
Antibodies
Type: Primary
Antigen: TEX19
Clonality: Monoclonal
Clone: 678826
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR