Taq DNA Polymerase with THERMOPOL® Buffer, New England Biolabs
Supplier: New England Biolabs (NEB)
M0267E M0267E M0267X M0267L M0267S
102877-582EA 2230.96 CAD
102877-582 CA102877-582 CA200067-186 CA101227-636 CA101227-638
Taq DNA Polymerase with THERMOPOL® Buffer, New England Biolabs
Nucleic Acid Reagents End-point PCR Enzymes and Kits
Taq DNA Polymerase is a thermostable DNA polymerase that possesses a 5´→ 3´ polymerase activity and a 5´ flap endonuclease activity.
It is the most widely used enzyme for PCR. To accomodate a variety of PCR applications, Taq is available with different reaction buffers. Standard Taq Reaction Buffer is designed to support existing PCR platforms, and is an ideal choice for DHPLC and high-throughput applications. ThermoPol Reaction Buffer was designed at NEB, and is formulated to promote high product yields, even under demanding conditions. For additional convenience, Taq DNA Polymerase is also available in kit and master mix formats. For direct gel loading, a Quick-Load version of the Taq 2X Master Mix is also available.