Omniprobe 3 Post, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
75964-08
102773-586EA 402.66 CAD
102773-586
Omniprobe 3 Post, Electron Microscopy Sciences
Graticules, Reticles and Grids
The 3 post copper lift-out grids are specifically designed for in-situ lift-out. These grids include multiple indexed mounting locations with both vertical bar and 'V' shaped attachment surfaces. These grids have a shallower downset and slightly wider center post.
Packaging: Packaged in glass vials.