Variable Tilt Mount holder, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
75952-05H 75952-05
102098-302EA 203.19 CAD
102098-302 102681-262
Variable Tilt Mount holder, Electron Microscopy Sciences
Specimen Mounting Supplies
This mount which is made from aluminum is used for the mounting and tilting of samples from 0° – 90° as well as for SEM investigation with small working distances. The mount is marked for 30°, 45°, 70° and 90°. The table measures 11 x 11 mm (0.46” x0.46”)and the overall size is 14 x 14 x 12.7 mm.